This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other POLE3 SIRNA Assays ›
Gene Symbol:
Pole3 LOC100362333
Gene Name:
polymerase (DNA directed), epsilon 3, accessory subunit
DNA-directed DNA polymerase epsilon 3
Gene Aliases:
-
-
Chromosome Location:
Chr.5: 78380813 - 78384047 on Build Rnor_6.0
Chromosome Location:
Chr.7: 118816052 - 118816729 on Build Rnor_6.0
UniGene ID:
Rn.3290
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID s151157

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pole3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007652.2 NP_001007653.1 4 325
GenBank mRNA BC081988.1 - 4 325 7129121
BC083800.1 - 4 325 7108773
Gene Symbol: LOC100362333
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_002729829.4 XP_002729875.1 1 386

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
histone H3 acetylation
DNA biosynthetic process
molecular_function
DNA-directed DNA polymerase activity
sequence-specific DNA binding
protein heterodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0