This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TM7SF3 SIRNA Assays ›
Gene Symbol:
Tm7sf3
Gene Name:
transmembrane 7 superfamily member 3
Gene Aliases:
-
Chromosome Location:
Chr.4: 180913703 - 180951564 on Build Rnor_6.0
UniGene ID:
Rn.23905
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s150863

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tm7sf3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011970.1 NP_001011970.1 12 1652
GenBank mRNA BC090030.1 - 12 1652 7366240

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_026281, AK171111, AK010720, AK154633, BC057900, and others.

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0