This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ATP6V1E1 SIRNA Assays ›
Gene Symbol:
Atp6v1e1
Gene Name:
ATPase, H+ transporting, lysosomal V1 subunit E1
Gene Aliases:
Atp6e, Atp6e2, P31, Vma4
Chromosome Location:
Chr.4: 153351434 - 153373558 on Build Rnor_6.0
UniGene ID:
Rn.103171
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s150777

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Atp6v1e1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198745.2 NP_942040.1 8 675
GenBank mRNA BC059155.1 - 8 679 6887702
FQ212928.1 - 8 675
FQ212966.1 - 8 670
FQ213001.1 - 8 675
FQ213103.1 - 8 672
FQ213345.1 - 8 677
FQ213479.1 - 8 676
FQ213531.1 - 8 677
FQ213667.1 - 8 694
FQ213669.1 - 8 677
FQ213903.1 - 8 672
FQ214254.1 - 8 677
FQ214381.1 - 8 675
FQ214440.1 - 8 677
FQ214497.1 - 8 643
FQ214920.1 - 8 675
FQ216629.1 - 8 675
FQ224865.1 - 8 676
FQ226034.1 - 8 670
FQ229770.1 - 2 83
FQ230006.1 - 8 675
FQ230733.1 - 8 670
FQ233079.1 - 8 675
FQ233569.1 - 8 677

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

metabolic process
ATP hydrolysis coupled proton transport
hydrogen-exporting ATPase activity, phosphorylative mechanism
proton-transporting ATPase activity, rotational mechanism
ATPase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0