siRNA Details
Gene Symbol: Dguok | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106602.1 | NP_001100072.1 | 3, 4 | 506 | ||
XM_006236735.2 | XP_006236797.1 | 4, 5 | 703 | |||
XM_006236736.2 | XP_006236798.1 | 4, 5 | 707 | |||
XM_006236737.2 | XP_006236799.1 | 3, 4 | 532 | |||
XM_008763056.1 | XP_008761278.1 | 4, 5 | 707 | |||
XM_008763057.1 | XP_008761279.1 | 3, 4 | 532 | |||
XM_008763058.1 | XP_008761280.1 | 4, 5 | 464 | |||
GenBank mRNA | BC168743.1 | - | 3, 4 | 447 | 6919887 |
