This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DGUOK SIRNA Assays ›
Gene Symbol:
Dguok
Gene Name:
deoxyguanosine kinase
Gene Aliases:
-
Chromosome Location:
Chr.4: 115180433 - 115208094 on Build Rnor_6.0
UniGene ID:
Rn.6770
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s150654

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dguok
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106602.1 NP_001100072.1 5 745
XM_006236735.2 XP_006236797.1 6 942
XM_006236736.2 XP_006236798.1 6 946
XM_006236737.2 XP_006236799.1 5 771
XM_008763056.1 XP_008761278.1 6 946
XM_008763057.1 XP_008761279.1 5 771
XM_008763058.1 XP_008761280.1 6 703
GenBank mRNA BC168743.1 - 5 686 6919887

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001162521, NM_013764, U90524, AY037862, AK167071, and others.

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
deoxyribonucleoside monophosphate biosynthetic process
nucleotide biosynthetic process
negative regulation of neuron projection development
dGTP metabolic process
purine deoxyribonucleoside metabolic process
molecular_function
deoxyguanosine kinase activity
ATP binding
nucleoside kinase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0