This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MTURN SIRNA Assays ›
Gene Symbol:
Mturn
Gene Name:
maturin, neural progenitor differentiation regulator homolog (Xenopus)
Gene Aliases:
-
Chromosome Location:
Chr.4: 84854353 - 84875300 on Build Rnor_6.0
UniGene ID:
Rn.92940
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s150592

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mturn
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007645.1 NP_001007646.1 2, 3 322
XM_008762880.1 XP_008761102.1 2, 3 355
GenBank mRNA BC083875.1 - 2, 3 322 7129817

Back To Top

More Information


Gene Ontology Categories:

Function(s)

multicellular organism development

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-6889b944d9-flx6z:80/100.66.128.142:80.
git-commit: df865406a845caddbd23a531a4b79e504db49c75
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header