This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FASTK SIRNA Assays ›
Gene Symbol:
Fastk
Gene Name:
Fas-activated serine/threonine kinase
Gene Aliases:
-
Chromosome Location:
Chr.4: 7260529 - 7264590 on Build Rnor_6.0
UniGene ID:
Rn.7680
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s150424

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fastk
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006235892.2 XP_006235954.1 6 1201
XM_006235894.2 XP_006235956.1 6 1188
XM_006235895.1 XP_006235957.1 6 1185
XM_006235896.2 XP_006235958.1 6 1202
GenBank mRNA BC083715.1 - 6 1205 7190690
FQ215763.1 - 7 1063
FQ228972.1 - 6 1141

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
regulation of RNA splicing
protein serine/threonine kinase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0