This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MAPKAP1 SIRNA Assays ›
Gene Symbol:
Mapkap1
Gene Name:
mitogen-activated protein kinase associated protein 1
Gene Aliases:
-
Chromosome Location:
Chr.3: 13484952 - 13701772 on Build Rnor_6.0
UniGene ID:
Rn.105886
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s150328

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mapkap1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011964.1 NP_001011964.1 4, 5 948
XM_006233949.2 XP_006234011.1 5, 6 1134
XM_006233950.2 XP_006234012.1 4, 5 686
XM_008761720.1 XP_008759942.1 4, 5 733
XM_008761721.1 XP_008759943.1 3, 4 648
XM_008761722.1 XP_008759944.1 2, 3 558
GenBank mRNA BC079073.1 - 4, 5 948 7122012

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_177345, BC090644, AK042154, AK052045, AK178100, and others.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
substantia nigra development
negative regulation of Ras protein signal transduction
molecular_function
phosphatidylinositol-4,5-bisphosphate binding
phosphatidylinositol-3,4,5-trisphosphate binding
Ras GTPase binding
protein kinase binding
phosphatidylinositol-3,4-bisphosphate binding
phosphatidic acid binding
phosphatidylinositol-3,5-bisphosphate binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0