This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF708 SIRNA Assays ›
Gene Symbol:
ZNF708
Gene Name:
zinc finger protein 708
Gene Aliases:
KOX8, ZNF15, ZNF15L1
Chromosome Location:
Chr.19: 21290388 - 21329439 on Build GRCh38
UniGene ID:
Hs.466296
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s15030

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF708
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001297560.1 NP_001284489.1 4 1237
NM_001297561.1 NP_001284490.1 3 1028
NM_021269.2 NP_067092.2 4 1153
XM_017027201.1 XP_016882690.1 4 1055
XM_017027202.1 XP_016882691.1 4 1178
XM_017027203.1 XP_016882692.1 2 958
XM_017027204.1 XP_016882693.1 4 970
GenBank mRNA AK128184.1 - 4 1112
AK131527.1 - 4 1153
BX647526.1 - 4 1223

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
RNA polymerase II core promoter proximal region sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0