This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TUBB4B SIRNA Assays ›
Gene Symbol:
Tubb4b
Gene Name:
tubulin, beta 4B class IVb
Gene Aliases:
Tubb2, Tubb2c
Chromosome Location:
Chr.3: 2441587 - 2444361 on Build Rnor_6.0
UniGene ID:
Rn.110562
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s150229

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tubb4b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199094.2 NP_954525.1 2 212
XM_006233582.2 XP_006233644.1 2 225
GenBank mRNA BC060597.1 - 2 216 6889737
FQ213825.1 - 2 209
FQ215022.1 - 2 209
FQ227178.1 - 2 209
FQ228801.1 - 2 209

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_177987, BC101271, BC101270.
This siRNA also targets the following transcripts in Mouse: NM_146116, AK146587, BC005547, AK006924, BC083319, and others.

Gene Ontology Categories:

Function(s) Process(es)

microtubule-based process
metabolic process
spindle assembly
protein polymerization
double-stranded RNA binding
GTPase activity
structural constituent of cytoskeleton
GTP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0