This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF711 SIRNA Assays ›
Gene Symbol:
ZNF711
Gene Name:
zinc finger protein 711
Gene Aliases:
CMPX1, MRX97, ZNF4, ZNF5, ZNF6, Zfp711, dJ75N13.1
Chromosome Location:
Chr.X: 85243820 - 85273362 on Build GRCh38
UniGene ID:
Hs.326801
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s15010

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 357.00
Your Price
Online offer: 296.31
296.3
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF711
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021998.4 NP_068838.3 4 903
XM_005262186.2 XP_005262243.1 5 1328
XM_005262187.1 XP_005262244.1 5 1217
XM_005262188.1 XP_005262245.1 4 903
XM_005262189.2 XP_005262246.1 5 1328
XM_011531019.1 XP_011529321.1 4 1703
XM_011531020.1 XP_011529322.1 5 1340
XM_011531021.1 XP_011529323.1 5 1345
XM_011531022.1 XP_011529324.1 5 1229
XM_011531023.2 XP_011529325.1 4 793
XM_011531024.1 XP_011529326.1 4 1691
XM_011531025.1 XP_011529327.1 4 1703
XM_011531026.1 XP_011529328.1 4 1691
XM_017029803.1 XP_016885292.1 2 1653
XM_017029804.1 XP_016885293.1 5 1333
XM_017029805.1 XP_016885294.1 3 783
XM_017029806.1 XP_016885295.1 2 1641
XM_017029807.1 XP_016885296.1 5 1333
XM_017029808.1 XP_016885297.1 5 1217
XM_017029809.1 XP_016885298.1 3 783
GenBank mRNA AK308097.1 - 5 1018
BC006349.2 - 4 862
BC067294.1 - 2 587 30386278
BX648117.1 - 4 1442

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
positive regulation of transcription, DNA-templated
RNA polymerase II regulatory region sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0