This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other METTL15 SIRNA Assays ›
Gene Symbol:
Mettl15
Gene Name:
methyltransferase like 15
Gene Aliases:
RGD1304978
Chromosome Location:
Chr.3: 100186106 - 100366148 on Build Rnor_6.0
UniGene ID:
Rn.160141
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s149746

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mettl15
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024981.1 NP_001020152.1 5 682
XM_006234686.2 XP_006234748.1 6 854
XM_006234687.2 XP_006234749.1 5 753
XM_006234688.2 XP_006234750.1 6 849
XM_006234689.2 XP_006234751.1 6 828
XM_008762084.1 XP_008760306.1 6 937
XM_008762085.1 XP_008760307.1 6 934
XM_008762086.1 XP_008760308.1 6 916
XM_008762087.1 XP_008760309.1 6 933
GenBank mRNA BC091215.1 - 5 682 7383613

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

rRNA base methylation
rRNA (cytosine-N4-)-methyltransferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0