This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other OLR528 SIRNA Assays ›
Gene Symbol:
Olr528 Olr522
Gene Name:
olfactory receptor 528
olfactory receptor 522
Gene Aliases:
-
ratchr3-70010454-70009507_ORF
Chromosome Location:
Chr.3: 74474561 - 74475502 on Build Rnor_6.0
Chromosome Location:
Chr.3: 74255120 - 74256067 on Build Rnor_6.0
UniGene ID:
Rn.142928
Rn.127929
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID s149550

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Olr528
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000319.1 NP_001000319.1 1 594
Gene Symbol: Olr522
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000562.1 NP_001000562.1 1 594

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
detection of chemical stimulus involved in sensory perception of smell
G-protein coupled receptor activity
olfactory receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0