This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other OLR495 SIRNA Assays ›
Gene Symbol:
Olr495 LOC100909940
Gene Name:
olfactory receptor 495
olfactory receptor 8J3-like
Gene Aliases:
-
-
Chromosome Location:
Chr.3: 73668052 - 73668999 on Build Rnor_6.0
Chromosome Location:
Chr.3: 74781470 - 74782417 on Build Rnor_6.0
UniGene ID:
Rn.142947
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID s149525

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Olr495
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000310.1 NP_001000310.1 1 907
Gene Symbol: LOC100909940
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003749508.2 XP_003749556.1 1 907

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_207563, NM_147078.

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
detection of chemical stimulus involved in sensory perception of smell
olfactory receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0