This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KCNMB2 SIRNA Assays ›
Gene Symbol:
Kcnmb2
Gene Name:
potassium channel subfamily M regulatory beta subunit 2
Gene Aliases:
Kcmb2
Chromosome Location:
Chr.2: 118547190 - 118585157 on Build Rnor_6.0
UniGene ID:
Rn.220612
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s148886

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Kcnmb2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_176861.1 NP_789831.1 3, 4 430
GenBank mRNA AB193523.1 - 1, 2 63
AJ517198.1 - 2, 3 314
AY191836.1 - 3, 4 430
BC081695.1 - 730 7190722

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_028231, AK014106, AK012400, AB193521, BC046227, and others.

Gene Ontology Categories:

Function(s) Process(es)

action potential
detection of calcium ion
potassium ion transport
chemical synaptic transmission
neuronal action potential
potassium ion transmembrane transport
potassium channel activity
calcium-activated potassium channel activity
potassium channel regulator activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0