This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PHF1 SIRNA Assays ›
Gene Symbol:
Phf1
Gene Name:
PHD finger protein 1
Gene Aliases:
Tctex3
Chromosome Location:
Chr.20: 5526928 - 5531940 on Build Rnor_6.0
UniGene ID:
Rn.24149
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s148467

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Phf1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_212538.1 NP_997703.1 14, 15 1620
XM_006256117.2 XP_006256179.1 14, 15 1874
XM_008772723.1 XP_008770945.1 15 1901
GenBank mRNA BC083843.1 - 14, 15 1550 7114670

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cellular response to DNA damage stimulus
biological_process
negative regulation of histone H3-K27 methylation
positive regulation of histone H3-K27 methylation
chromatin binding
protein binding
zinc ion binding
methylated histone binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0