This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other OLR1690 SIRNA Assays ›
Gene Symbol:
Olr1690 LOC100910382 LOC100910696 Olr1688
Gene Name:
olfactory receptor 1690
olfactory receptor 14J1-like
olfactory receptor 14J1-like
olfactory receptor 1688
Gene Aliases:
-
-
-
ratchr20-540075-541079_ORF
Chromosome Location:
Chr.20: 775501 - 776472 on Build Rnor_6.0
Chromosome Location:
Chr.20: 734629 - 735681 on Build Rnor_6.0
Chromosome Location:
Chr.20: 955239 - 956260 on Build Rnor_6.0
Chromosome Location:
Chr.20: 915079 - 916083 on Build Rnor_6.0
UniGene ID:
Rn.142687
-
-
Rn.142689
Species:
Rat
Species Specific ID (Flybase ID):
-
-
-
-
siRNA ID s148337

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Olr1690
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000274.1 NP_001000274.1 1 61
Gene Symbol: LOC100910382
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003751905.2 XP_003751953.1 1 74
Gene Symbol: LOC100910696
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008772684.1 XP_008770906.1 84
Gene Symbol: Olr1688
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000275.1 NP_001000275.1 61

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
detection of chemical stimulus involved in sensory perception of smell
G-protein coupled receptor activity
olfactory receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0