This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other BRMS1 SIRNA Assays ›
Gene Symbol:
Brms1 LOC100911841
Gene Name:
breast cancer metastasis-suppressor 1
breast cancer metastasis-suppressor 1 homolog
Gene Aliases:
-
-
Chromosome Location:
Chr.1: 220418516 - 220427780 on Build Rnor_6.0
Chromosome Location:
Chr.1: 220325292 - 220334621 on Build Rnor_6.0
UniGene ID:
Rn.2839
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID s147801

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Brms1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009605.1 NP_001009605.1 470
GenBank mRNA BC088469.1 - 470 7128111
Gene Symbol: LOC100911841
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003749038.3 XP_003749086.1 5 530

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
apoptotic process
histone deacetylation
negative regulation of NF-kappaB transcription factor activity
negative regulation of transcription, DNA-templated
positive regulation of protein deacetylation
positive regulation of anoikis
cellular process
histone deacetylase activity
histone deacetylase binding
NF-kappaB binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0