This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other IRF7 SIRNA Assays ›
Gene Symbol:
Irf7
Gene Name:
interferon regulatory factor 7
Gene Aliases:
-
Chromosome Location:
Chr.1: 214249388 - 214252909 on Build Rnor_6.0
UniGene ID:
Rn.101159
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s147730

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Irf7
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033691.1 NP_001028863.1 9 1721
XM_006230540.1 XP_006230602.1 10 1820
XM_006230541.1 XP_006230603.1 10 1724
GenBank mRNA BC104686.1 - 9 1721 7442863
FQ232130.1 - 8 1176
FQ232859.1 - 7 1013

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of adaptive immune response
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
immunoglobulin mediated immune response
interferon-alpha production
interferon-beta production
positive regulation of interferon-alpha production
regulation of MyD88-dependent toll-like receptor signaling pathway
regulation of MyD88-independent toll-like receptor signaling pathway
type I interferon biosynthetic process
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of type I interferon-mediated signaling pathway
RNA polymerase II core promoter sequence-specific DNA binding
transcription factor activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0