This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KIF22 SIRNA Assays ›
Gene Symbol:
Kif22
Gene Name:
kinesin family member 22
Gene Aliases:
-
Chromosome Location:
Chr.1: 198461406 - 198476430 on Build Rnor_6.0
UniGene ID:
Rn.8601
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s147587

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Kif22
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009645.1 NP_001009645.1 9 1294
GenBank mRNA BC088421.1 - 9 1294 7123493

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_145588, BC003427, AK075922, AK084310, AK088033.

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
microtubule-based movement
sister chromatid cohesion
mitotic metaphase plate congression
metabolic process
DNA binding
microtubule motor activity
ATP binding
microtubule binding
ATPase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0