This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other OLR229 SIRNA Assays ›
Gene Symbol:
Olr229 LOC100911839 Olr234
Gene Name:
olfactory receptor 229
olfactory receptor 2D3-like
olfactory receptor 234
Gene Aliases:
-
-
-
Chromosome Location:
Chr.1: 170693928 - 170694872 on Build Rnor_6.0
Chromosome Location:
Chr.1: 171161968 - 171163055 on Build Rnor_6.0
Chromosome Location:
Chr.1: 171274305 - 171275249 on Build Rnor_6.0
UniGene ID:
Rn.141789
-
Rn.141785
Species:
Rat
Species Specific ID (Flybase ID):
-
-
-
siRNA ID s147419

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Olr229
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000204.1 NP_001000204.1 55
Gene Symbol: LOC100911839
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006229964.2 XP_006230026.1 1 198
Gene Symbol: Olr234
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000208.1 NP_001000208.1 1 55

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
detection of chemical stimulus involved in sensory perception of smell
olfactory receptor activity
G-protein coupled receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0