This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other UQCRH SIRNA Assays ›
Gene Symbol:
UQCRH
Gene Name:
ubiquinol-cytochrome c reductase hinge protein
Gene Aliases:
QCR6, UQCR8
Chromosome Location:
Chr.1: 46303613 - 46316777 on Build GRCh38
UniGene ID:
Hs.481571
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s14709

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 357.00
Your Price
Online offer: 296.31
296.3
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: UQCRH
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001297565.1 NP_001284494.1 4 479
NM_001297566.1 NP_001284495.1 2 304
NM_006004.3 NP_005995.2 3 331
GenBank mRNA AK311967.1 - 3 227
BC001426.2 - 3 231 3140323
BC001934.1 - 3 236 2823237
BC015177.1 - 3 234 3905169
BC093060.1 - 4 394 30409772
BC107703.1 - 3 237 6605146
BG472687.1 - 2 205
BP366403.1 - 4 481
BT007070.1 - 3 177

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

oxidative phosphorylation
mitochondrial electron transport, ubiquinol to cytochrome c
aerobic respiration
oxidation-reduction process
hydrogen ion transmembrane transport
protein binding
ubiquinol-cytochrome-c reductase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0