This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other L7RN6 SIRNA Assays ›
Gene Symbol:
l7Rn6
Gene Name:
lethal, Chr 7, Rinchik 6
Gene Aliases:
-
Chromosome Location:
Chr.1: 154147098 - 154170428 on Build Rnor_6.0
UniGene ID:
Rn.96145
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s147030

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: l7Rn6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013897.1 NP_001013919.1 5, 6 778
XM_006229662.2 XP_006229724.1 4, 5 745
XM_008759638.1 XP_008757860.1 4, 5 576
XM_008759639.1 XP_008757861.1 3, 4 505
GenBank mRNA BC088340.1 - 5, 6 778 7367791
FQ212130.1 - 4, 5 689
FQ213824.1 - 4, 5 690
FQ214677.1 - 4, 5 690
FQ215506.1 - 4, 5 681
FQ216197.1 - 4, 5 690
FQ216668.1 - 4, 5 690
FQ216759.1 - 4, 5 690
FQ218863.1 - 4, 5 687
FQ220044.1 - 4, 5 690
FQ220310.1 - 4, 5 741
FQ220575.1 - 4, 5 690
FQ224272.1 - 4, 5 692
FQ224945.1 - 4, 5 690
FQ229506.1 - 4, 5 686

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_016401, AK223198, AF161487, BC018080, BC015991, and others.
This siRNA also targets the following transcripts in Mouse: NM_026304, AK012166, AK002306, AK003324, BC083330, and others.

Gene Ontology Categories:

Function(s) Process(es)

protein import into nucleus
Golgi organization
protein transport
lung development
cellular response to heat
protein transporter activity
Hsp70 protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0