This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CPEB1 SIRNA Assays ›
Gene Symbol:
Cpeb1
Gene Name:
cytoplasmic polyadenylation element binding protein 1
Gene Aliases:
-
Chromosome Location:
Chr.1: 143171457 - 143278485 on Build Rnor_6.0
UniGene ID:
Rn.224485
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s146981

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cpeb1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106276.1 NP_001099746.1 3, 4 416
XM_008759509.1 XP_008757731.1 3, 4 1025
XM_008759510.1 XP_008757732.1 3, 4 1025

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_007755, NM_001252525, NM_001252526, BY720025, BC144948, and others.

Gene Ontology Categories:

Function(s) Process(es)

mRNA processing
regulation of translation
synaptonemal complex assembly
negative regulation of cell proliferation
positive regulation of neuron projection development
negative regulation of translation
positive regulation of cell migration
cellular response to insulin stimulus
positive regulation of translation
regulation of neuronal synaptic plasticity
mRNA transport
positive regulation of nitric-oxide synthase biosynthetic process
cellular response to lipopolysaccharide
cellular response to amino acid stimulus
cellular response to hypoxia
negative regulation of cytoplasmic translation
nucleotide binding
translation repressor activity, nucleic acid binding
RNA binding
mRNA binding
mRNA 3'-UTR binding
mRNA 3'-UTR AU-rich region binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0