This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KLHL25 SIRNA Assays ›
Gene Symbol:
Klhl25
Gene Name:
kelch-like family member 25
Gene Aliases:
RGD1310815
Chromosome Location:
Chr.1: 137325459 - 137359104 on Build Rnor_6.0
UniGene ID:
Rn.16545
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s146946

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Klhl25
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039006.1 NP_001034095.1 2 562
XM_006229379.2 XP_006229441.1 2 875
XM_006229380.2 XP_006229442.1 2 4913
XM_006229381.2 XP_006229443.1 2 3968
XM_008759505.1 XP_008757727.1 2 605
GenBank mRNA BC099111.1 - 2 562 7461562
FQ230538.1 - 2 570

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_029652, NM_001122780, NM_182782, BC027373, AK201755, and others.

Gene Ontology Categories:

Function(s)

regulation of translational initiation
protein ubiquitination
protein ubiquitination involved in ubiquitin-dependent protein catabolic process

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0