This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GLTSCR2 SIRNA Assays ›
Gene Symbol:
Gltscr2
Gene Name:
glioma tumor suppressor candidate region gene 2
Gene Aliases:
-
Chromosome Location:
Chr.1: 77836522 - 77843991 on Build Rnor_6.0
UniGene ID:
Rn.8075
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s146569

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gltscr2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207591.1 NP_997474.1 3 358
GenBank mRNA AY535002.1 - 3 358
BC086359.1 - 3 362 7304013
FQ212888.1 - 3 389
FQ213201.1 - 3 368
FQ218937.1 - 3 364
FQ219058.1 - 3 364
FQ222789.1 - 3 368

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_133831, AK151836, BY283382, AY535001, AK169290, and others.

Gene Ontology Categories:

Process(es)

poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0