This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TDRD1 SIRNA Assays ›
Gene Symbol:
Tdrd1
Gene Name:
tudor domain containing 1
Gene Aliases:
-
Chromosome Location:
Chr.1: 277637474 - 277685236 on Build Rnor_6.0
UniGene ID:
Rn.156064
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s146299

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tdrd1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106202.1 NP_001099672.1 14 1926
XM_006231642.2 XP_006231704.1 15 2189
XM_008760524.1 XP_008758746.1 15 1996
XM_008760525.1 XP_008758747.1 15 1961
XM_008760526.1 XP_008758748.1 14 2126
XM_008760528.1 XP_008758750.1 14 1755
XM_008760529.1 XP_008758751.1 14 2033
XM_008760530.1 XP_008758752.1 15 2189

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001002241, NM_001002240, NM_031387, NM_001002238, AF285591, and others.

Gene Ontology Categories:

Function(s) Process(es)

germ cell development
spermatogenesis
gene silencing by RNA
piRNA metabolic process
DNA methylation involved in gamete generation
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0