This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SF3B3 SIRNA Assays ›
Gene Symbol:
Sf3b3
Gene Name:
splicing factor 3b, subunit 3
Gene Aliases:
-
Chromosome Location:
Chr.19: 43392067 - 43429582 on Build Rnor_6.0
UniGene ID:
Rn.7554
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s146201

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Sf3b3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106187.2 NP_001099657.2 17 2317
XM_006255620.2 XP_006255682.1 17 2452
GenBank mRNA BC168883.1 - 11 1222

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

mRNA splicing, via spliceosome
RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0