This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DSC2 SIRNA Assays ›
Gene Symbol:
Dsc2
Gene Name:
desmocollin 2
Gene Aliases:
-
Chromosome Location:
Chr.18: 11826705 - 11858808 on Build Rnor_6.0
UniGene ID:
Rn.3954
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s145943

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dsc2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033688.1 NP_001028860.1 10 1501
XM_008771935.1 XP_008770157.1 10 1565
GenBank mRNA BC101864.1 - 10 1501 7939139

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_013505, L33779, AK172168, X73885, BC004663, and others.

Gene Ontology Categories:

Function(s) Process(es)

homophilic cell adhesion via plasma membrane adhesion molecules
cellular response to starvation
ventricular cardiac muscle cell action potential
cardiac muscle cell-cardiac muscle cell adhesion
bundle of His cell to Purkinje myocyte communication
regulation of heart rate by cardiac conduction
calcium ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0