This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KCTD16 SIRNA Assays ›
Gene Symbol:
Kctd16
Gene Name:
potassium channel tetramerization domain containing 16
Gene Aliases:
RGD1559856
Chromosome Location:
Chr.18: 33445437 - 33727382 on Build Rnor_6.0
UniGene ID:
Rn.38640
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s145812

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Kctd16
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001172155.1 NP_001165626.1 1 753
GenBank mRNA FQ212362.1 - 1731

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_026135, AK220381.

Gene Ontology Categories:

Function(s)

regulation of G-protein coupled receptor protein signaling pathway
protein homooligomerization

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0