This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SKA1 SIRNA Assays ›
Gene Symbol:
Ska1
Gene Name:
spindle and kinetochore associated complex subunit 1
Gene Aliases:
RGD1310784
Chromosome Location:
Chr.18: 70174316 - 70184368 on Build Rnor_6.0
UniGene ID:
Rn.106906
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s145691

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ska1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106134.1 NP_001099604.1 6 663
XM_006254918.2 XP_006254980.1 6 676
XM_006254920.2 XP_006254982.1 6 669
XM_008772106.1 XP_008770328.1 6 751
XM_008772107.1 XP_008770329.1 6 747
XM_008772108.1 XP_008770330.1 6 749
GenBank mRNA BC158659.1 - 6 615 8369180

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

chromosome segregation
mitotic nuclear division
regulation of microtubule polymerization or depolymerization
cell division
microtubule binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0