This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CRLF1 SIRNA Assays ›
Gene Symbol:
Crlf1
Gene Name:
cytokine receptor-like factor 1
Gene Aliases:
-
Chromosome Location:
Chr.16: 20675042 - 20686360 on Build Rnor_6.0
UniGene ID:
Rn.38637
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s145129

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Crlf1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106074.1 NP_001099544.1 6 1065
XM_006252872.2 XP_006252934.1 6 1108
XM_006252874.2 XP_006252936.1 6 1108

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

ureteric bud development
biological_process
positive regulation of cell proliferation
positive regulation of tyrosine phosphorylation of Stat3 protein
negative regulation of neuron apoptotic process
negative regulation of motor neuron apoptotic process
molecular_function
cytokine activity
ciliary neurotrophic factor receptor binding
cytokine binding
protein heterodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0