This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CHDH SIRNA Assays ›
Gene Symbol:
Chdh
Gene Name:
choline dehydrogenase
Gene Aliases:
CDH, CHD
Chromosome Location:
Chr.16: 6078086 - 6109386 on Build Rnor_6.0
UniGene ID:
Rn.22857
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s145009

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Chdh
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198731.2 NP_942026.1 8 1950
XM_006252578.2 XP_006252640.1 8 2015
XM_006252579.2 XP_006252641.1 8 1969
XM_006252580.2 XP_006252642.1 8 1775
XM_008770984.1 XP_008769206.1 8 2318
XM_008770985.1 XP_008769207.1 9 2407
GenBank mRNA AY365023.1 - 7 1699
BC085787.1 - 8 1950 7108667
X94769.1 - 7 1225

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

glycine betaine biosynthetic process from choline
oxidation-reduction process
choline dehydrogenase activity
flavin adenine dinucleotide binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0