siRNA Details
Gene Symbol: Pla2g3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106015.1 | NP_001099485.1 | 2 | 663 |

Gene Symbol: Pla2g3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106015.1 | NP_001099485.1 | 2 | 663 |