This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TSPYL1 SIRNA Assays ›
Gene Symbol:
TSPYL1
Gene Name:
TSPY like 1
Gene Aliases:
TSPYL
Chromosome Location:
Chr.6: 116274859 - 116280117 on Build GRCh38
UniGene ID:
Hs.458358
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s14456

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TSPYL1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003309.3 NP_003300.1 1 1505
GenBank mRNA AF042181.1 - 1 128
AK026849.1 - 1 366
AL136629.1 - 1 1319
BC048969.1 - 1 1321 5297724
BC068554.1 - 1 1318
CR533535.1 - 1 1221

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

nucleosome assembly
biological_process
enzyme binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0