This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FIP1L1 SIRNA Assays ›
Gene Symbol:
Fip1l1
Gene Name:
factor interacting with PAPOLA and CPSF1
Gene Aliases:
-
Chromosome Location:
Chr.14: 36147864 - 36208384 on Build Rnor_6.0
UniGene ID:
Rn.138993
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s144463

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fip1l1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008295.1 NP_001008296.1 8 739
XM_006250882.2 XP_006250944.1 8 843
XM_006250883.2 XP_006250945.1 8 843
XM_006250884.2 XP_006250946.1 8 841
XM_006250885.2 XP_006250947.1 8 841
XM_006250886.2 XP_006250948.1 8 845
XM_006250887.2 XP_006250949.1 8 839
GenBank mRNA BC085767.1 - 8 739 7192362
EU224276.1 - 8 738

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

mRNA processing
biological_process
molecular_function
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0