This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ANKRD17 SIRNA Assays ›
Gene Symbol:
Ankrd17
Gene Name:
ankyrin repeat domain 17
Gene Aliases:
RGD1562348
Chromosome Location:
Chr.14: 19293635 - 19431521 on Build Rnor_6.0
UniGene ID:
Rn.8442
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s144422

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ankrd17
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105999.1 NP_001099469.1 12 2575
XM_006250763.2 XP_006250825.1 12 2112
XM_006250764.2 XP_006250826.1 12 2112
XM_006250765.2 XP_006250827.1 12 2058
XM_006250766.2 XP_006250828.1 12 2694
XM_006250767.2 XP_006250829.1 12 2994
XM_006250768.2 XP_006250830.1 12 2112
XM_006250769.2 XP_006250831.1 12 2112
XM_006250770.2 XP_006250832.1 12 2058
XM_008770011.1 XP_008768233.1 12 2694
GenBank mRNA FQ235158.1 - 3 194

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

blood vessel maturation
regulation of DNA replication
defense response to bacterium
positive regulation of I-kappaB kinase/NF-kappaB signaling
innate immune response
positive regulation of cell cycle
negative regulation of smooth muscle cell differentiation
positive regulation of G1/S transition of mitotic cell cycle
positive regulation of MDA-5 signaling pathway
positive regulation of RIG-I signaling pathway
chromatin binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0