This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RGD1311892 SIRNA Assays ›
Gene Symbol:
RGD1311892
Gene Name:
similar to hypothetical protein FLJ10901
Gene Aliases:
-
Chromosome Location:
Chr.13: 53088438 - 53108713 on Build Rnor_6.0
UniGene ID:
Rn.48092
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s144338

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RGD1311892
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134502.1 NP_001127974.1 2 477
XM_006249920.2 XP_006249982.1 2 338
XM_006249922.2 XP_006249984.1 2 624

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_028872, AK017848, BB611110, BC053100, BC052416.

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header