This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RAB3GAP2 SIRNA Assays ›
Gene Symbol:
Rab3gap2
Gene Name:
RAB3 GTPase activating protein subunit 2
Gene Aliases:
RGD1311518
Chromosome Location:
Chr.13: 103157806 - 103230750 on Build Rnor_6.0
UniGene ID:
Rn.101730
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s144296

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rab3gap2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040154.2 NP_001035244.2 32 3836
XM_008769835.1 33 3821
XM_008769836.1 XP_008768058.1 31 5242
XM_008769837.1 XP_008768059.1 30 3512
GenBank mRNA BC086380.1 - 6 677

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of GTPase activity
positive regulation of GTPase activity
establishment of protein localization to endoplasmic reticulum membrane
positive regulation of endoplasmic reticulum tubular network organization
GTPase activator activity
Rab guanyl-nucleotide exchange factor activity
Rab GTPase binding
enzyme regulator activity
protein heterodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0