This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TRPS1 SIRNA Assays ›
Gene Symbol:
TRPS1
Gene Name:
transcriptional repressor GATA binding 1
Gene Aliases:
GC79, LGCR
Chromosome Location:
Chr.8: 115408496 - 115669633 on Build GRCh38
UniGene ID:
Hs.657018
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s14429

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TRPS1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282902.2 NP_001269831.1 6 3146
NM_001282903.2 NP_001269832.1 7 3635
NM_014112.4 NP_054831.2 7 3649
XM_005251049.2 XP_005251106.1 6 3464
XM_006716625.1 XP_006716688.1 7 3586
XM_011517264.1 XP_011515566.1 8 3719
XM_011517266.2 XP_011515568.1 7 3454
XM_011517267.1 XP_011515569.1 7 3572
XM_011517268.1 XP_011515570.1 7 3561
XM_017013801.1 XP_016869290.1 6 7248
GenBank mRNA AF183810.1 - 7 3643
AF264784.1 - 6 3190
AK000948.1 - 4 1633
AK304046.1 - 6 3141
BC125020.1 - 7 3479 40073132

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
skeletal system development
transcription from RNA polymerase II promoter
NLS-bearing protein import into nucleus
heart development
organ morphogenesis
tissue development
regulation of chondrocyte differentiation
cell fate commitment
positive regulation of transcription from RNA polymerase II promoter
cell development
digestive tract development
anatomical structure formation involved in morphogenesis
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II transcription factor binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
protein binding
zinc ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-fd98957d6-rbs6c:80/100.66.128.142:80.
git-commit: 278a916a254c4ca07f9ccbb2b9efa8cd7578b5be
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0