This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DISP1 SIRNA Assays ›
Gene Symbol:
Disp1
Gene Name:
dispatched homolog 1 (Drosophila)
Gene Aliases:
-
Chromosome Location:
Chr.13: 101451932 - 101597570 on Build Rnor_6.0
UniGene ID:
Rn.41748
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s144288

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Disp1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105983.1 NP_001099453.1 10 2877
XM_006250381.2 XP_006250443.1 9 2746
XM_006250383.2 XP_006250445.1 8 3207
XM_006250384.2 XP_006250446.1 8 2686
XM_008769827.1 XP_008768049.1 11 3571
XM_008769828.1 XP_008768050.1 9 2857

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

smoothened signaling pathway
patched ligand maturation
determination of left/right symmetry
pattern specification process
biological_process
embryonic pattern specification
dorsal/ventral pattern formation
peptide transport
diaphragm development
molecular_function
hedgehog receptor activity
peptide transporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header