This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TRPS1 SIRNA Assays ›
Gene Symbol:
TRPS1
Gene Name:
transcriptional repressor GATA binding 1
Gene Aliases:
GC79, LGCR
Chromosome Location:
Chr.8: 115408496 - 115669633 on Build GRCh38
UniGene ID:
Hs.657018
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s14427

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 357.00
Your Price
Online offer: 296.31
296.3
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TRPS1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282902.2 NP_001269831.1 3 1808
NM_001282903.2 NP_001269832.1 4 2297
NM_014112.4 NP_054831.2 4 2311
XM_005251049.2 XP_005251106.1 3 2126
XM_006716625.1 XP_006716688.1 4 2248
XM_011517264.1 XP_011515566.1 5 2381
XM_011517266.2 XP_011515568.1 4 2116
XM_011517267.1 XP_011515569.1 4 2234
XM_011517268.1 XP_011515570.1 4 2223
XM_017013801.1 XP_016869290.1 3 5910
GenBank mRNA AF183810.1 - 4 2305
AF264784.1 - 3 1852
AK000948.1 - 1 295
AK304046.1 - 3 1803
AK309357.1 - 3 2022
BC125020.1 - 4 2141 40073132

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
skeletal system development
transcription from RNA polymerase II promoter
NLS-bearing protein import into nucleus
heart development
organ morphogenesis
tissue development
regulation of chondrocyte differentiation
cell fate commitment
positive regulation of transcription from RNA polymerase II promoter
cell development
digestive tract development
anatomical structure formation involved in morphogenesis
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II transcription factor binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
protein binding
zinc ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header