This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DESI2 SIRNA Assays ›
Gene Symbol:
Desi2
Gene Name:
desumoylating isopeptidase 2
Gene Aliases:
CGI-146, DeSI-2, Fam152a, PNAS-4, Pppde1
Chromosome Location:
Chr.13: 96111189 - 96156693 on Build Rnor_6.0
UniGene ID:
Rn.9401
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s144235

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Desi2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013873.1 NP_001013895.1 3 546
XM_006250316.2 XP_006250378.1 3 896
XM_006250317.2 XP_006250379.1 4 1310
XM_008769783.1 XP_008768005.1 3 853
XM_008769784.1 XP_008768006.1 4 1029
XM_008769785.1 XP_008768007.1 3 1157
GenBank mRNA BC083584.1 - 3 546 7131107
FQ226068.1 - 3 321

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

proteolysis
peptidase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0