This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RGD1564093 SIRNA Assays ›
Gene Symbol:
RGD1564093 LOC501934
Gene Name:
similar to RIKEN cDNA 2310036O22
uncharacterized protein C19orf43 homolog
Gene Aliases:
-
-
Chromosome Location:
Chr.19: 26142720 - 26146136 on Build Rnor_6.0
Chromosome Location:
Chr.X: 116557457 - 116558148 on Build Rnor_6.0
UniGene ID:
Rn.121465
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID s143955

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RGD1564093
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105946.1 NP_001099416.1 2, 3 483
GenBank mRNA FQ231178.1 - 2, 3 422
FQ235293.1 - 2, 3 422
Gene Symbol: LOC501934
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008773502.1 XP_008771724.1 2 159

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_024038, BC004533, AF061732, BC015347, AK027588, and others.
This siRNA also targets the following transcripts in Mouse: NM_026760, AK003299, BC023284, AK009653, BC029622.

Panther Classification:

Molecular Function -

DNA metabolism protein

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0