This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other OLR1000 SIRNA Assays ›
Gene Symbol:
Olr1000 Olr959 Olr960
Gene Name:
olfactory receptor 1000
olfactory receptor 959
olfactory receptor 960
Gene Aliases:
ratchr7-6430339-6429404_ORF
-
-
Chromosome Location:
Chr.7: 5700094 - 5701028 on Build Rnor_6.0
Chromosome Location:
Chr.7: 6970616 - 6971557 on Build Rnor_6.0
Chromosome Location:
Chr.7: 6690738 - 6691679 on Build Rnor_6.0
UniGene ID:
Rn.143368
Rn.225571
Rn.141026
Species:
Rat
Species Specific ID (Flybase ID):
-
-
-
siRNA ID s143917

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Olr1000
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000077.1 NP_001000077.1 1 514
Gene Symbol: Olr959
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000486.1 NP_001000486.1 1 514
Gene Symbol: Olr960
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000589.1 NP_001000589.1 514

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
detection of chemical stimulus involved in sensory perception of smell
G-protein coupled receptor activity
olfactory receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0