This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other OLR903 SIRNA Assays ›
Gene Symbol:
Olr903 LOC103689923
Gene Name:
olfactory receptor 903
olfactory receptor 6C2-like
Gene Aliases:
-
-
Chromosome Location:
Chr.7: 5401508 - 5402443 on Build Rnor_6.0
Chromosome Location:
Chr.7: 3900228 - 3901163 on Build Rnor_6.0
UniGene ID:
Rn.143396
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID s143825

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Olr903
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001361.1 NP_001001361.1 1 70
Gene Symbol: LOC103689923
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008765069.1 XP_008763291.1 1 70

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
detection of chemical stimulus involved in sensory perception of smell
G-protein coupled receptor activity
olfactory receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7bff9b55f5-ncxmj:80/100.66.128.142:80.
git-commit: 5f36da1a8ad9bc9722227bfd86ebc827ae27d083
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0