This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TMEM248 SIRNA Assays ›
Gene Symbol:
Tmem248
Gene Name:
transmembrane protein 248
Gene Aliases:
-
Chromosome Location:
Chr.12: 29931728 - 29958050 on Build Rnor_6.0
UniGene ID:
Rn.99691
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s143637

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tmem248
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004204.2 NP_001004204.1 8, 9 1341
XM_006249230.2 XP_006249292.1 5, 6 1043
XM_006249231.2 XP_006249293.1 5, 6 964
GenBank mRNA BC079161.1 - 5, 6 1015 7129411
BC090318.1 - 8, 9 1341 7113430

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0