This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TMEM120A SIRNA Assays ›
Gene Symbol:
Tmem120a
Gene Name:
transmembrane protein 120A
Gene Aliases:
RGD1311474, Tmpit
Chromosome Location:
Chr.12: 23989596 - 23998257 on Build Rnor_6.0
UniGene ID:
Rn.6288
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s143608

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tmem120a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010945.1 NP_001010945.1 3 622
GenBank mRNA BC089063.1 - 3 622 7310299

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_172541, BC046757, AK217162, AK028119.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0