This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FIS1 SIRNA Assays ›
Gene Symbol:
Fis1
Gene Name:
fission 1 (mitochondrial outer membrane) homolog (S. cerevisiae)
Gene Aliases:
Ttc11
Chromosome Location:
Chr.12: 22750485 - 22765315 on Build Rnor_6.0
UniGene ID:
Rn.1298
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s143594

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fis1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006249121.2 XP_006249183.1 1, 2 377
XM_006249123.2 XP_006249185.1 1, 2 357
GenBank mRNA FQ213833.1 - 1, 2 137
FQ222246.1 - 1, 2 123
FQ224250.1 - 1, 2 134
FQ232488.1 - 1, 2 123

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_025562, AK008323, AK008916, AK008078, AK004917, and others.

Gene Ontology Categories:

Function(s) Process(es)

mitochondrial fission
mitophagy
release of cytochrome c from mitochondria
protein targeting to mitochondrion
positive regulation of cytosolic calcium ion concentration
mitochondrial fusion
regulation of mitochondrion organization
peroxisome fission
negative regulation of endoplasmic reticulum calcium ion concentration
calcium-mediated signaling using intracellular calcium source
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
mitochondrial fragmentation involved in apoptotic process
protein homooligomerization
positive regulation of mitochondrial calcium ion concentration
mitochondrion morphogenesis
positive regulation of mitochondrial fission
positive regulation of protein targeting to membrane
positive regulation of intrinsic apoptotic signaling pathway
receptor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0