This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other HCLS1 SIRNA Assays ›
Gene Symbol:
Hcls1
Gene Name:
hematopoietic cell specific Lyn substrate 1
Gene Aliases:
-
Chromosome Location:
Chr.11: 66736966 - 66759402 on Build Rnor_6.0
UniGene ID:
Rn.15578
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s143212

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Hcls1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011898.1 NP_001011898.1 7 534
GenBank mRNA BC079081.1 - 7 534 7122756

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008225, BC007469, X84797, D42120, AK088723.

Panther Classification:

Molecular Function -

actin or actin-binding cytoskeletal protein cytoskeletal protein non-motor actin binding protein

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
positive regulation of cell proliferation
response to hormone
positive regulation of phosphatidylinositol 3-kinase signaling
actin filament polymerization
erythrocyte differentiation
regulation of actin filament polymerization
positive regulation of granulocyte differentiation
positive regulation of peptidyl-serine phosphorylation
positive regulation of tyrosine phosphorylation of STAT protein
positive regulation of transcription factor import into nucleus
positive regulation of macrophage differentiation
positive regulation of transcription from RNA polymerase II promoter
positive regulation of peptidyl-tyrosine phosphorylation
positive regulation of protein kinase B signaling
cellular response to cytokine stimulus
negative regulation of leukocyte apoptotic process
RNA polymerase II transcription factor binding
actin binding
SH3 domain binding
protein kinase binding
protein complex binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0